Learn More
VITLAB™ Class B Polypropylene Volumetric Flask
Set volumes accurately for exact measurements in lab applications with these flasks. VITLAB™ Class B Polypropylene Volumetric Flasks feature a single calibration mark that defines the calibrated volume exactly.
Marke:  VITLAB™ 671941
17.13 CHF gültig bis 2025-12-31
Benutzen Sie den Promotions-Code "25339" um Ihren Promotions-Preis zu erhalten.
Artikelnummer. 11883652
  • 17.60 CHF / 2 Stück
Nur noch null auf Lager
Verfügbar ab: 09-10-2025
Beschreibung
  • Highly transparent
  • With ring mark individually calibrated to ‘In’
  • Class B tolerances according to DIN EN ISO 1042
  • Thermal stress up to 60°C does not permanently exceed the tolerance limits
  • To preserve markings, cleaning at no higher than 60°C is recommended
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
VITLAB™ Class B Polypropylene Volumetric Flask > Capacity: 25mL; Height: 115mm; Includes: NS 10/19 stopper; Tolerance: ±0.08mL