missing translation for 'onlineSavingsMsg'
Learn More

Phenomenex™ Claricep™ Empty Screw-On Flash Columns with Snap Lock and 2 Frits

Artikelnummer. 31748973 Alle ansehen Phenomenex Produkte
Ansicht ändern
Click to view available options
Säulenbettgewicht (metrisch):
12 g
80 g
120 g
220 g
330 g
800 g
1.5 kg
3 kg
5 kg
Packungsgröße:
1 Stück
20 Stück
5 Stück
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Säulenbettgewicht (metrisch) Packungsgröße
31748973 80 g 5 Stück
31744533 12 g 20 Stück
31741561 120 g 5 Stück
31742892 1.5 kg 1 Stück
31746839 220 g 1 Stück
31747800 3 kg 1 Stück
31743636 330 g 1 Stück
31742993 5 kg 1 Stück
31741927 800 g 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 31748973 Supplier Phenomenex™ Supplier No. FCH080H
Verfügbar ab: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Available in two flash media (irregular and spherical) with multiple chemistries, Claricep columns are optimized for resolution and high loading capacity. The deactivated silica surface promotes compound stability, excellent peak shape and high performance for both acidic and basic compounds.

  • Brand: Claricep
  • Technique: Accessory
TRUSTED_SUSTAINABILITY

Specifications

Typ Empty Cartridge
Säulenbettgewicht (metrisch) 80 g
Menge 5 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.