missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CK1 delta Monoclonal Antibody (C3), Invitrogen™

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA550957
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CSNK1D (CK1 delta), a serine/threonine kinase, plays a critical role in Wnt/beta-catenin signaling pathway. This pathway is important in both development and cancer. The encoded protein may also be involved in the regulation of apoptosis, circadian rhythm, microtubule dynamics, chromosome segregation, and p53-mediated effects on growth. The encoded protein is highly similar to the mouse and rat CK1 delta homologs. Three transcript variants encoding different isoforms have been found for CK1 delta.
Spezifikation
| CK1 delta | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P48730, Q06486 | |
| CSNK1D | |
| Recombinant Human CSNK1d protein, Met1-Lys409 (Accession #P48730). | |
| 200 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C3 | |
| Unconjugated | |
| CSNK1D | |
| 1200006A05Rik; AA409348; ASPS; casein kinase 1; casein kinase 1 delta; casein kinase 1, delta; casein kinase I isoform delta; CK1d; CK1delta; CKId; CKIdelta; CKI-delta; CSNK1D; D930010H05Rik; FASPS2; HCKID; HGNC:2452; Tau-protein kinase CSNK1D | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100625051, 1453, 64462 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur