missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chromogranin C Polyclonal Antibody

Goat Polyclonal Antibody
Marke: Invitrogen™ PA5143108
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Studies in rodents suggest that the full-length protein, secretogranin II, is involved in the packaging or sorting of peptide hormones and neuropeptides into secretory vesicles. The full-length protein is cleaved to produce the active peptide secretoneurin, which exerts chemotaxic effects on specific cell types, and EM66, whose function is unknown.
Spezifikation
| Chromogranin C | |
| Polyclonal | |
| Unconjugated | |
| SCG2 | |
| Chcg; Chgc; Chromogranin C (Secretogranin II); chromogranin-C; EM66; Manserin; SCG2; Scg-2; secretogranin 2; secretogranin II; secretogranin II (chromogranin C); secretogranin-2; Secretoneurin; SgII; SN | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 7857 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P13521 | |
| SCG2 | |
| Peptide with sequence C-QELGKLTGPNNQKRE. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur