Learn More
Chromogranin C Polyclonal Antibody
Chromogranin C Polyclonal Antibody
Marke: Thermo Scientific PA5115018
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Chromogranin C Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Spezifikation
| Chromogranin C | |
| Polyclonal | |
| Unconjugated | |
| SCG2 | |
| Chcg; Chgc; Chromogranin C (Secretogranin II); chromogranin-C; EM66; Manserin; SCG2; Scg-2; secretogranin 2; secretogranin II; secretogranin II (chromogranin C); secretogranin-2; Secretoneurin; SgII; SN | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20254, 24765, 7857 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P10362, P13521, Q03517 | |
| SCG2 | |
| A synthesized peptide derived from human SCG2(Accession P13521), corresponding to amino acid residues K49-L99. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |