missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chromogranin C Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA584972
This item is not returnable.
View return policy
Description
Immunogen sequence: NPVEEKIESQ TQEEVRDSKE NIEKNEQIND EMKRSGQLGI QEEDLRKESK DQLSDDVSKV IAYLKRLVNA AGSGRLQNGQ NGERA Highest antigen sequence indentity to the following orthologs: Rat - 62%, Mouse - 76%.
The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Studies in rodents suggest that the full-length protein, secretogranin II, is involved in the packaging or sorting of peptide hormones and neuropeptides into secretory vesicles. The full-length protein is cleaved to produce the active peptide secretoneurin, which exerts chemotaxic effects on specific cell types, and EM66, whose function is unknown.
Specifications
| Chromogranin C | |
| Polyclonal | |
| Unconjugated | |
| SCG2 | |
| Chcg; Chgc; Chromogranin C (Secretogranin II); chromogranin-C; EM66; Manserin; SCG2; Scg-2; secretogranin 2; secretogranin II; secretogranin II (chromogranin C); secretogranin-2; Secretoneurin; SgII; SN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7857 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P13521 | |
| SCG2 | |
| Recombinant protein corresponding to Human SCG2. Recombinant protein control fragment (Product #RP-108272). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction