missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chromogranin C Polyclonal Antibody
Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA530718
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Recommended positive controls: IMR32, U87-MG, mouse brain, rat brain. Predicted reactivity: Mouse (89%), Rat (88%), Pig (88%), Rhesus Monkey (96%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Studies in rodents suggest that the full-length protein, secretogranin II, is involved in the packaging or sorting of peptide hormones and neuropeptides into secretory vesicles. The full-length protein is cleaved to produce the active peptide secretoneurin, which exerts chemotaxic effects on specific cell types, and EM66, whose function is unknown.
Spezifikation
| Chromogranin C | |
| Polyclonal | |
| Unconjugated | |
| SCG2 | |
| Chcg; Chgc; Chromogranin C (Secretogranin II); chromogranin-C; EM66; Manserin; SCG2; Scg-2; secretogranin 2; secretogranin II; secretogranin II (chromogranin C); secretogranin-2; Secretoneurin; SgII; SN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20254, 7857 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.26 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P13521, Q03517 | |
| SCG2 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human Chromogranin C. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur