missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chromogranin B Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA552605
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: KQYSSHHTAE KRKRLGELFN PYYDPLQWKS SHFERRDNMN DNFLEGEEEN ELTLNEKNFF PEYNYDWWEK KPFSEDVNWG YEKRNLARVP KLDLKRQYDR VAQLDQLLHY RKKSAEFPDF YDS Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.
Chromogranin B is a tyrosine sulfated secretory protein found in a wide variety of peptidergic endocrine cells. Chromogranin functions as a neuroendocrine secretory granule protein which may be the precursor for other biologically active peptides.
Spezifikation
| Chromogranin B | |
| Polyclonal | |
| Unconjugated | |
| Chgb | |
| CCB peptide; CCB peptide long form; CCB peptide short form; CgB; CHGB; chromogranin B; chromogranin B (secretogranin 1); Chromogranin B, parathyroid secretory protein; chromogranin-B; GAWK peptide; Glucagonoma peptide; PE-11; SCG1; Scg-1; secretogranin B; secretogranin I; Secretogranin-1; secretogranin-I; SgI | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1114 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P05060 | |
| Chgb | |
| Recombinant protein corresponding to Human Chromogranin B. Recombinant protein control fragment (Product #RP-89888). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur