Learn More
Chromogranin A / CHGA (Neuroendocrine Marker) Mouse, Clone: rCHGA/798, enQuireBio™
Mouse Monoclonal Antibody
234.00 CHF - 449.92 CHF
Spezifikation
| Antigen | Chromogranin A / CHGA (Neuroendocrine Marker) |
|---|---|
| Klon | rCHGA/798 |
| Konzentration | 200μg/mL |
| Anwendungen | Immunohistochemistry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15979777
|
EnquireBio
1113-MSM17-P0 |
20 μg | |||||||
|
15989777
|
EnquireBio
1113-MSM17-P1 |
100 μg |
549.15 CHF 449.92 CHF / 100 Mikrogramm Sparen 99.23 CHF 18% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Spezifikation
| Chromogranin A / CHGA (Neuroendocrine Marker) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human CHGA protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 68-75kDa |
| rCHGA/798 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 1113 | |
| IgG1 κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Finely granular cytoplasmic | |
| PC12 cells. Adrenal gland, bowel, thyroid, pancreas, or pheochromocytoma. |