missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11553-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome.Spezifikation
CHRNB1 | |
Polyclonal | |
Unconjugated | |
CHRNB1 | |
ACHRB, CHRNB, CHRNB1, CMS1D, CMS2A, SCCMS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1140, 11443, 24261 | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09690, P11230, P25109 | |
CHRNB1 | |
CHRNB1 Fusion Protein Ag2108 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Liquid |