Learn More
Chicken anti-Human IgG (H+L) Secondary Antibody, DyLight 550, Novus Biologicals™
Chicken Polyclonal Antibody
Marke:  Novus Biologicals NB120-6713R
Artikelnummer. 18742453
  • 435.00 CHF / 0.50 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
IgG (H+L)
Polyclonal
50mM Sodium Borate
Human IgG whole molecule
0.5 mL
Adaptive Immunity, Immunology
Human
IgG
Western Blot, ELISA, Immunohistochemistry, Dot Blot
DyLight 550
Chicken
Multi-step
RUO
Secondary
Store at 4°C in the dark.
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen