missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP57 (19-128) Mouse anti-Human, Clone: 1E9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CEP57.
Marke: Abnova H00009702-M02.100ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Translokin binds basic fibroblast growth factor (FGF2; MIM 134920) and mediates its nuclear translocation and mitogenic activity (Bossard et al., 2003 [PubMed 12717444]).[supplied by OMIM]
Sequence: AEPSRSNGSMVRHSSSPYVVYPSDKPFLNSDLRRSPSKPTLAYPESNSRAIFSALKNLQDKIRRLELERIQAEESVKTLSRETIEYKKVLDEQIQERENSKNEESKHNQESpezifikation
| CEP57 (19-128) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014679 | |
| CEP57 | |
| CEP57 (NP_055494,19 a.a. ∽ 128 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Antibody Reactive Against Recombinant Protein. | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| ELISA, Immunofluorescence, Western Blot | |
| 1E9 | |
| Mouse monoclonal antibody raised against a partial recombinant CEP57. | |
| CEP57 | |
| KIAA0092/PIG8/TSP57 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 9702 | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction