Beschreibung
- Very compact footprint to fit on every lab bench
- Very quiet operation to benefit your environment
- Low access height for simple loading and unloading of samples
- SOFT brake function for slow acceleration and braking. Optimized for cell separation by gradient centrifugation
- 'At set RPM' function starts the timer when the selected rotational speed is reached for reproducible centrifugation runs
- Key lock to prevent unintentional adjustments
- Stainless steel rotor chamber is rust-free and easy to clean
- Electronic imbalance detection for maximum safety
Spezifikation
Spezifikation
| Fassungsvermögen | 4 x 100 mL |
| Max. RZB | 3,000 x g |
| Max. Geschwindigkeit | 4,400 rpm |
| Gekühlt | No |
| Reglertyp | Knobs |
| Rotor inklusive | No |
| Produkttyp | Benchtop Centrifuge , Switzerland IVD Version |