Beschreibung
- Remarkable versatility with 12 different rotors
- Soft-touch one-finger lid closure for ergonomic operation
- Menu-driven, multi lingual operation menu (English, German, French, Spanish) with large backlit display
- 5 program keys for easy access to routine programs
- Saves up to 50 user-defined programs
- Automatic rotor recognition and imbalance detection for maximized operational safety
- Eppendorf QuickLock™ - System for quickly opening and closing the rotor lid
Spezifikation
Spezifikation
| Fassungsvermögen | 30 x 1.5/2 mL Tubes |
| Max. Geschwindigkeit | 14,000 rpm |
| Max. RZB | 20,817 x g |
| Reglertyp | Knobs |
| Rotor inklusive | Yes |
| Gekühlt | Yes |
| Rotor | FA-45-30-11 |
| Produkttyp | Microcentrifuge, IVD Switzerland Version |