Description
- Remarkable versatility with 12 different rotors
- Soft-touch one-finger lid closure for ergonomic operation
- Menu-driven, multi lingual operation menu (English, German, French, Spanish) with large backlit display
- 5 program keys for easy access to routine programs
- Saves up to 50 user-defined programs
- Automatic rotor recognition and imbalance detection for maximized operational safety
- Eppendorf QuickLock™ - System for quickly opening and closing the rotor lid
Specifications
Specifications
| Fassungsvermögen | 48 x 1.5/2.0 mL, (6 x 15/50 mL, 2 x MTP) |
| Max. Geschwindigkeit | 17,500 rpm |
| Max. RZB | 30,130 x g |
| Reglertyp | Knobs |
| Rotor inklusive | No |
| Gekühlt | No |
| Produkttyp | Microcentrifuge, IVD Switzerland Version |