Beschreibung
- Soft-touch one-finger lid closure for ergonomic lid locking
- Automatic lid opening at the end of the run to prevent sample warming and to allow easy access to samples
- High quality anodized aluminum rotors and rotor lids
- One push short-spin key for quick centrifugation - no need to hold key all the while
- Easy to clean keypad control panel
- Particularly small footprint to save precious space on the lab bench
Spezifikation
Spezifikation
| Fassungsvermögen | 24 x 1.5/2 mL Tubes |
| Max. Geschwindigkeit | 15,060 rpm |
| Max. RZB | 21,300 x g |
| Reglertyp | Keypad |
| Rotor inklusive | No |
| Gekühlt | Yes |
| Produktlinie | Eppendorf 5425 R |
| Produkttyp | Microcentrifuge; IVD Switzerland Version |