Learn More
Eppendorf™ Centrifuge 5420 - Microcentrifuge
BIOPHARMA
Artikelnummer. 30689768 Alle ansehen Eppendorf Produkte
Ansicht ändern
Click to view available options
Rotor inklusive:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Rotor inklusive
30689768 Yes
30689748 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30689768 Lieferant Eppendorf™ Lieferanten-Nr. 5420000105
 Eingeschränkter Artikel
Verfügbar ab: 15-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
The Eppendorf Centrifuge 5420 offers 24-place capacity in a small footprint and supports nucleic acid purification and gel-extraction applications.
  • Soft-touch one-finger lid closure for ergonomic lid locking
  • Automatic lid opening at the end of the run to prevent sample warming and to allow easy access to samples
  • High quality anodized aluminum rotors and rotor lids
  • One push short-spin key for quick centrifugation - no need to hold key all the while
  • Easy to clean keypad control panel
  • Particularly small footprint to save precious space on the lab bench
TRUSTED_SUSTAINABILITY
Tekniske data
Fassungsvermögen 24 x 1.5/2 mL Tubes
Max. Geschwindigkeit 15,060 rpm
Max. RZB 21,300 x g
Rotor inklusive Yes
Gekühlt No
Produkttyp Microcentrifuge; IVD Switzerland Version
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.