missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPJ Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11517-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein that belongs to the centromere protein family. During cell division, this protein plays a structural role in the maintece of centrosome integrity and normal spindle morphology, and it is involved in microtubule disassembly at the centrosome. This protein can function as a transcriptional coactivator in the Stat5 signaling pathway, and also as a coactivator of NF-kappaB-mediated transcription, likely via its interaction with the coactivator p300/CREB-binding protein. Mutations in this gene are associated with primary autosomal recessive microcephaly, a disorder characterized by severely reduced brain size and mental retardation.Spezifikation
CENPJ | |
Polyclonal | |
Unconjugated | |
CENPJ | |
BM032, CENP J, CENPJ, centromere protein J, CPAP, LAG 3 associated protein, LAP, LIP1, LYST interacting protein 1, MCPH6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
219103, 305909, 55835 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q569L8, Q9HC77 | |
CENPJ | |
CENPJ Fusion Protein Ag2078 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |