Learn More
Celiprolol Hydrochloride, TRC
Artikelnummer. 30132864
Change view
Click to view available options
Menge:
5 mg
10 mg
100 mg
Packungsgröße:
10 Milligramm
100 Milligramm
5 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30132864 100 mg 100 Milligramm
17927464 10 mg 10 Milligramm
17909664 5 mg 5 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132864 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCC254500100MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 57470-78-7
Summenformel C20 H33 N3 O4 . Cl H
Molekulargewicht (g/mol) 415.95
Synonym Celiprolol Hydrochloride, Celectol, Corliprol, NSC 324509, REV 5320A, ST 1396, Selecor, Urea, N'-[3-acetyl-4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-N,N-diethyl-, monohydrochloride (9CI)
IUPAC-Name 3-[3-acetyl-4-[3-(tert-butylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea;hydrochloride
SMILES Cl.CCN(CC)C(=O)Nc1ccc(OCC(O)CNC(C)(C)C)c(c1)C(=O)C
Spezifikation
Chemischer Name oder Material Celiprolol Hydrochloride
Hinweise zur Reinheitsqualität HPLC
Menge 100 mg
InChI-Formel InChI=1S/C20H33N3O4.ClH/c1-7-23(8-2)19(26)22-15-9-10-18(17(11-15)14(3)24)27-13-16(25)12-21-20(4,5)6;/h9-11,16,21,25H,7-8,12-13H2,1-6H3,(H,22,26);1H
Formelmasse 415.2238
Reinheit (%) >95%