missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Ceftazidime Hydrate
Click to view available options
Menge:
1 g
5 g
Packungsgröße:
1 Gramm
5 Gramm
Beschreibung
Ceftazidime is a third-generation cephalosporin antibiotic used in research settings. It is effective against a broad spectrum of bacteria, including Gram-negative and some Gram-positive bacteria.
Ceftazidime works by inhibiting bacterial cell wall synthesis, ultimately leading to the death of susceptible bacteria. It may be utilized as a selective agent in microbiology and molecular biology experiments. Incorporate ceftazidime into culture media to selectively promote the growth of bacterial strains that carry resistance genes against this antibiotic. This is often achieved by introducing a plasmid or vector containing a ceftazidime resistance gene into the bacteria of interest.
Spezifikation
Spezifikation
| Chemischer Name oder Material | Ceftazidime Hydrate |
| CAS | 72558-82-8 |
| Menge | 5 g |
| Summenformel | C22H22N6O7S2 - xH2O |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur