missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEBPB Mouse anti-Human, Rat, Clone: 2B6E10, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66649-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.Spezifikation
CEBPB | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P17676, P21272 | |
CEBPB | |
CEBPB Fusion Protein Ag20073 | |
150 μL | |
Primary | |
Human, Rat | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2B6E10 | |
Unconjugated | |
CEBPB | |
C/EBP beta, CEBPB, CRP2, IL6DBP, LAP, Liver activator protein, NF IL6, Nuclear factor NF IL6, TCF 5, TCF5, Transcription factor 5 | |
Mouse | |
Protein G | |
RUO | |
1051, 24253 | |
Antibody | |
IgG1 |