missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDH26 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20057-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody can recognize all the isoforms of CDH26.
Cadherins are a family of adhesion molecules that mediate Ca2+-dependent cell-cell adhesion in all solid tissues and modulate a wide variety of processes, including cell polarization and migration. Cadherin domains occur as repeats in the extracellular region and are thought to contribute to the sorting of heterogeneous cell types and the maintece of orderly structures such as epithelium. This gene encodes a cadherin domain-containing protein whose specific function has not yet been determined. Alternative splicing occurs at this locus and two transcript variants, encoding distinct proteins, have been identified.Spezifikation
CDH26 | |
Polyclonal | |
Unconjugated | |
CDH26 | |
cadherin like 26, Cadherin like protein 26, Cadherin like protein VR20, CDH26, VR20 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
60437 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IXH8 | |
CDH26 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |