missing translation for 'onlineSavingsMsg'
Learn More

CDC42SE2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 21424-1-AP-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16841025

  • 381.34 CHF / 150 Mikroliter
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Probably involved in the organization of the actin cytoskeleton by acting downstream of CDC42, inducing actin filament assembly. Alters CDC42-induced cell shape changes. In activated T-cells, may play a role in CDC42-mediated F-actin accumulation at the immunological synapse. May play a role in early contractile events in phagocytosis in macrophages.
Spezifikation

Spezifikation

CDC42SE2
Polyclonal
Unconjugated
CDC42SE2
CDC42 small effector 2, CDC42 small effector protein 2, CDC42SE2, SPEC2
Rabbit
Antigen Affinity Chromatography
RUO
56990
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.24 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NRR3
CDC42SE2
CDC42SE2 Fusion Protein Ag16011
150 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote