missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90 Mouse anti-Human, Porcine, Rat, Clone: 2D7D11, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66766-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
THY1 encodes a cell surface glycoprotein and member of the immunoglobulin superfamily of proteins. The encoded protein is involved in cell adhesion and cell communication in numerous cell types, but particularly in cells of the immune and nervous systems. The encoded protein is widely used as a marker for hematopoietic stem cells. This gene may function as a tumor suppressor in nasopharyngeal carcinoma. Alternative splicing results in multiple transcript variants.Spezifikation
CD90 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01830, P04216 | |
THY1 | |
CD90 Fusion Protein Ag25603 | |
150 μL | |
Primary | |
Human, Porcine, Rat | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2D7D11 | |
Unconjugated | |
THY1 | |
CD90, CDw90, FLJ33325, Thy 1 antigen, Thy 1 cell surface antigen, Thy 1 membrane glycoprotein, THY1 | |
Mouse | |
Protein A | |
RUO | |
100271931, 24832, 7070 | |
Antibody | |
IgG2a |