missing translation for 'onlineSavingsMsg'
Learn More

CD90.1 (Thy-1.1) Mouse anti-Mouse, Rat, Super Bright 645, Clone: HIS51, eBioscience™

Mouse Monoclonal Antibody

Marke:  Invitrogen 64-0900-80

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983624

  • 144.00 CHF / 25 Mikrogramm


Verfügbar ab
05-10-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. It is expressed in murine T cells, thymocytes, neural cells, Kupffer′s cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain.
Spezifikation

Spezifikation

CD90.1 (Thy-1.1)
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
P01830, P01831
Thy1
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
21838, 24832
Flow Cytometry
HIS51
Super Bright 645
Thy1
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse, Rat
Videos
Dokumentation