Learn More
CD90.1 (Thy-1.1) Monoclonal Antibody (HIS51), Super Bright™ 645, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
393.00 CHF
Spezifikation
Antigen | CD90.1 (Thy-1.1) |
---|---|
Klon | HIS51 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Beschreibung
Description: The 53-2.1 monoclonal antibody reacts with mouse CD90.2 also known as Thy-1.2, a GPI-linked membrane molecule. CD90.2 is expressed by mouse thymocytes and mature T cells as well as neurons in CD90.2-expressing mouse strains. These strains include BALB/c, CBA, C3H, C57BL/6, C58/, SJL and others. Cells from CD90.1-expressing strains including PL and AKR do not stain with 53-2.1. CD90 is involved in regulation of adhesion and signal transduction by T cells. Applications Reported: This 53-2.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 53-2.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No.
CD90.1 (Thy-1.1) Monoclonal antibody specifically detects CD90.1 (Thy-1.1) in Mouse, Rat samples. It is validated for Flow Cytometry.Spezifikation
CD90.1 (Thy-1.1) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P01830, P01831 | |
Thy1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Thy1 |
HIS51 | |
Flow Cytometry | |
Super Bright 645 | |
Mouse | |
Mouse, Rat | |
21838, 24832 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |