Learn More
CD8a Mouse, PerCP-Cy5.5, Clone: RPA-T8, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio QAB19-PCP55-25Tests
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD8a | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD8A | |
Protein A/G Purified | |
RUO | |
925 | |
IgG1 κ |
Flow Cytometry | |
RPA-T8 | |
PerCP-Cyanine5.5 | |
NP_741969 | |
Mouse | |
25 Tests | |
Primary | |
Store at 2-4°C. |