missing translation for 'onlineSavingsMsg'
Learn More

CD8a Mouse anti-Human, Super Bright 702, Clone: RPA-T8, eBioscience™

Mouse Monoclonal Antibody

Marke:  Invitrogen 67-0088-41

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15825713

  • 208.00 CHF / 25 Stück


Verfügbar ab
02-06-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Description: The eBioCB-CALLA monoclonal antibody recognizes human CD10 (CALLA, NEP, enkephalinase, Neprilysin), which is a 100 kDa, type II cell surface glycoprotein originally identified for its expression on most acute lymphoblastic leukemias (ALL). Subsequently, CD10 was shown to be the same molecule as the neutral endopeptidase (NEP), or KII-NA. CD10 is a Zn2+-dependent metallo-peptidase with endothelin, glucagon, gastrin, neurotensin and bradykinin included among its substrates. CD10 is involved in the regulation of chemotactic and inflammatory processes involving neutrophils. In B cells, CD10 regulates stromal cell-dependent B lymphopoiesis and expression has also been reported on mature B cells in germinal centres. In addition to the hematopoietic compartment, other major sites of CD10 expression are the brush border of enterocytes and renal tubules and glomeruli. There is partial blocking of the eBioCB-CALLA and MEM-78 monoclonal antibodies indicating that they recognize similar epitopes. Applications Reported: This eBioCB-CALLA (CB-CALLA) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioCB-CALLA (CB-CALLA) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 702 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).
Spezifikation

Spezifikation

CD8a
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P01732
CD8A
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
925
Flow Cytometry
RPA-T8
Super Bright 702
CD8A
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only