missing translation for 'onlineSavingsMsg'
Learn More

CD8a, Super Bright 600, clone: RPA-T8, eBioscience™

Mouse Monoclonal Antibody

Marke:  Invitrogen 63-0088-41

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15805553

  • 180.00 CHF / 25 Stück


Geschätzter Ausliefertermin
von Anbieter 28-03-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Light sensitivity: This tandem dye is sensitive to photo- induced oxidation. Protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 2-8°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorochrome performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 600 nm; Laser: Violet Laser CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).
Spezifikation

Spezifikation

CD8a
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P01732
CD8 alpha, leu-2a
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
RPA-T8
Super Bright 600
CD8A
Liquid
CD8A
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
925
Videos

For Research Use Only