Learn More
CD8a Mouse anti-Human, CoraLite 488, Clone: RPA-T8, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-65144-100TESTS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The CD8a antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD8a. This antibody recognizes human antigen. The CD8a antibody has been validated for the following applications: FC analysis. This antibody is coralite™488.
Spezifikation
| CD8a | |
| Monoclonal | |
| 5 μL/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| RPA-T8 | |
| CoraLite 488 | |
| 100 Tests | |
| 925 | |
| IgG1 |