Learn More
CD8a Monoclonal Antibody (53-6.7), Super Bright™ 702, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 67-0081-80
Additional Details : Gewicht : 0.01000kg
Description
Description: The eBioH35-17.2 monoclonal antibody reacts with the mouse CD8 beta molecule. The CD8 beta chain associates with the CD8 alpha chain to form the CD8 alpha/beta heterodimer expressed on the surface of a majority of thymocytes, and on peripheral cytotoxic alpha beta TCR T cells. CD8 binds to MHC class I and plays a role in T cell development and activation of mature T cells. Applications Reported: This eBioH35-17.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioH35-17.2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400-42) to minimize any non-specific polymer interactions.
When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 702 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).Specifications
CD8a | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P01731 | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
53-6.7 | |
Super Bright 702 | |
Cd8a | |
CD8A | |
Affinity chromatography | |
RUO | |
12525 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
For Research Use Only