Learn More
CD86 (B7-2) Monoclonal Antibody (GL1), Super Bright™ 645, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
193.00 CHF - 388.00 CHF
Spezifikation
Antigen | CD86 (B7-2) |
---|---|
Klon | GL1 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15825673
|
Affymetrix eBioscience
64-0862-80 |
25 μg |
193.00 CHF
25 Mikrogramm |
Verfügbar ab: 10-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15835673
|
Affymetrix eBioscience
64-0862-82 |
100 μg |
388.00 CHF
100 Mikrogramm |
Verfügbar ab: 10-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Description: The IT2.2 monoclonal antibody reacts with human CD86, an ∽80 kDa surface receptor also known as B7-2. CD86 and CD80 are members of the B7 family of costimulatory molecules. CD86 is expressed at low levels on B cells, macrophages, and dendritic cells and is upregulated on B cells through a variety of surface stimuli including the BCR complex, CD40 and some cytokine receptors. In addition to CD80 (B7-1), CD86 is a counter-receptor for the T cell surface molecules CD28 and CD152 (CTLA-4). The interaction of CD86 with its ligands plays a critical role in T-B crosstalk, T cell costimulation, autoantibody production and Th2-mediated Ig production. The kinetics of upregulation of CD86 upon stimulation supports its major contribution during the primary phase of an immune response. Applications Reported: This IT2.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This IT2.2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter.
CD86 (B7-2) Monoclonal antibody specifically detects CD86 (B7-2) in Mouse samples. It is validated for Flow Cytometry.Spezifikation
CD86 (B7-2) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
activation B7-2 antigen; B lymphocyte activation antigen B72; B7; B7.2; B70; B7-2; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; Cd28l2; CD28LG2; CD86; CD86 antigen; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CD86 antigen precursor; CD86 molecule; CLS1; CTLA-4 counter-receptor B7.2; early T cell costimulatory molecule-1; early T-cell costimulatory molecule 1; Early T-cell co-stimulatory molecule 1; ETC-1; FUN-1; LAB72; Ly58; Ly-58; MB7; MB7-2; membrane glycoprotein; MGC34413; T-lymphocyte activation antigen CD86; TS/A-2 | |
CD86 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD86 |
GL1 | |
Flow Cytometry | |
Super Bright 645 | |
Rat | |
Mouse | |
P42082 | |
12524 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |