missing translation for 'onlineSavingsMsg'
Learn More

CD83, Super Bright 436, clone: HB15e, eBioscience™

Mouse Monoclonal Antibody

Marke:  Invitrogen 62-0839-41

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15805503

  • 188.00 CHF / 25 Stück


Geschätzter Ausliefertermin
von Anbieter 04-03-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Description: The HB15e monoclonal antibody reacts with human CD83, a 45kDa transmembrane glycoprotein. CD83, a member of the Ig superfamily, is expressed on cultured dendritic cells, interdigitating, follicular, and circulating dendritic cells as well as some proliferating lymphocytes, and human cell lines express this antigen. While the function of CD83 is unclear, it can serve as a useful marker for mature human blood dendritic cells.Applications Reported: This HB15e antibody has been reported for use in flow cytometric analysis.Applications Tested: This HB15e antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5μL (1.0μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 105 to 10 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser
Spezifikation

Spezifikation

CD83
Monoclonal
5μl/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
Q01151
CD83
IgG1, kappa
25 Tests
4°C, store in dark, DO NOT FREEZE!
Primary
9308
Flow Cytometry
HB15e
Super Bright 436
CD83
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos

For Research Use Only