missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79a Mouse, Clone: IGA/1406, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 973-MSM6-P0
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD79a | |
Cell surface | |
IGA/1406 | |
Unconjugated | |
NP_067612 | |
Mouse | |
25-44kDa | |
Protein A/G Purified | |
RUO | |
973 | |
IgG3 |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CD79A | |
A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein. | |
Daudi or Ramos cells. Germinal center B-cells in a lymph node or tonsil. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |