Learn More
CD79A Mouse anti-Human, Clone: IGA/515, Abnova™
Mouse monoclonal antibody raised against full length recombinant human CD79A.
Marke: Abnova MAB13228.100ug
Additional Details : Gewicht : 0.02000kg
Beschreibung
The B lymphocyte antigen receptor is a multimeric complex that includes the antigen-specific component, surface immunoglobulin (Ig). Surface Ig non-covalently associates with two other proteins, Ig-alpha and Ig-beta, which are necessary for expression and function of the B-cell antigen receptor. This gene encodes the Ig-alpha protein of the B-cell antigen component. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]
Spezifikation
CD79A | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
IGA/MB-1 | |
Mouse | |
Protein A/G purification | |
RUO | |
973 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
IGA/515 | |
Mouse monoclonal antibody raised against full length recombinant human CD79A. | |
In 10mM PBS. | |
CD79A | |
Recombinant protein corresponding to full length of human CD79A. | |
100 μg | |
Primary | |
Human | |
Liquid |