Learn More
CD62L (L-Selectin) Monoclonal Antibody (DREG-56 (DREG56)), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
146.00 CHF - 373.00 CHF
Spezifikation
Antigen | CD62L (L-Selectin) |
---|---|
Klon | DREG-56 (DREG56) |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15963384
|
Affymetrix eBioscience
62-0629-42 |
100 Tests |
373.00 CHF
100 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15953384
|
Affymetrix eBioscience
62-0629-41 |
25 Tests |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Description: The DREG-56 monoclonal antibody reacts with human CD62L, a 76 kDa member of the selectin family. CD62L is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells and binds a number of glycosylated, fucosylated, sulfated sialylated glycoproteins including CD34, glycam-1 and MAdCAM-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues. Applications Reported: This DREG-56 antibody has been reported for use in flow cytometric analysis. Applications Tested: This DREG-56 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated mouse lymph node cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions.
The human CD62L is a 74-95 kDa glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.Spezifikation
CD62L (L-Selectin) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P14151 | |
SELL | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
SELL |
DREG-56 (DREG56) | |
Flow Cytometry | |
Super Bright 436 | |
Mouse | |
Human | |
6402 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |