Learn More
CD56 Mouse anti-Human, PE-Cy5, Clone: B159, BD
Mouse Monoclonal Antibody
280.00 CHF - 545.00 CHF
Spezifikation
Antigen | CD56 |
---|---|
Klon | B159 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | PE-Cyanine5 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15891449
|
BD Biosciences
561904 |
25 Tests |
280.00 CHF
1 Stück |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15828238
|
BD Biosciences
555517 |
100 Tests |
545.00 CHF
1 Stück |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.
Spezifikation
CD56 | |
Flow Cytometry | |
PE-Cyanine5 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39 |
B159 | |
Monoclonal | |
Mouse | |
Human | |
Human NK Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.