missing translation for 'onlineSavingsMsg'
Learn More

CD56 Mouse anti-Human, Alexa Fluor 488, Clone: B159, BD

Mouse Monoclonal Antibody

204.00 CHF - 417.00 CHF

Spezifikation

Antigen CD56
Klon B159
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Konjugat Alexa Fluor 488
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15801459
Unterlagen ansehen Aktionsdetails
BD Biosciences
561905
25 Tests
204.00 CHF
1 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15828998
Unterlagen ansehen Aktionsdetails
BD Biosciences
557699
100 Tests
417.00 CHF
1 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Spezifikation

Spezifikation

CD56
Flow Cytometry
Alexa Fluor 488
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39
B159
Monoclonal
Mouse
Human
Human NK Cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

For Research Use Only.