missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Mouse, PerCP, Clone: L17F12, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio QAB14-PCP-500Tests
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD5 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD5 | |
Protein A/G Purified | |
RUO | |
921 | |
IgG2a κ |
Flow Cytometry | |
L17F12 | |
PerCP | |
NP_055022 | |
Mouse | |
500 Tests | |
Primary | |
Store at 2-4°C. |