missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Mouse anti-Human, APC, Clone: UCHT2, BD
Mouse Monoclonal Antibody
217.00 CHF - 565.00 CHF
Spezifikation
Antigen | CD5 |
---|---|
Klon | UCHT2 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | APC |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15830679
|
BD Biosciences
561003 |
25 Tests |
217.00 CHF
1 Stück |
Verfügbar ab: 27-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15898088
|
BD Biosciences
555355 |
100 Tests |
565.00 CHF
1 Stück |
Verfügbar ab: 27-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The UCHT2 monoclonal antibody specifically binds to CD5. CD5 is a 67 kDa single-chain, type 1 transmembrane glycoprotein expressed on most thymocytes, the majority of peripheral T lymphocytes and a subpopulation of B cells. CD72 has been shown to be the natural ligand for CD5. CD5+ B cells produce polyreactive antibodies (mostly IgM).
Spezifikation
CD5 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
UCHT2 | |
Monoclonal | |
Mouse | |
Human | |
Human T cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.