Learn More
CD5 Monoclonal Antibody (UCHT2), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
177.00 CHF - 425.00 CHF
Spezifikation
Antigen | CD5 |
---|---|
Klon | UCHT2 |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15805483
|
Affymetrix eBioscience
62-0059-42 |
100 Tests |
425.00 CHF
100 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15895473
|
Affymetrix eBioscience
62-0059-41 |
25 Tests |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Description: The UCHT2 monoclonal antibody reacts with human CD5, a 67 kDa protein expressed by a majority of thymocytes and mature T cells and a subset of B cells. Signaling through the CD5 molecule activates T cells and binding of CD5 to its ligand on B cells, CD72, and plays an important role in T-B interaction and proliferation. The monoclonal antibody UCHT2 recognizes primate CD5. Applications Reported: This UCHT2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This UCHT2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser
CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD5 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P06127 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
UCHT2 | |
Flow Cytometry | |
Super Bright 436 | |
Mouse | |
Human | |
921 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only