missing translation for 'onlineSavingsMsg'
Learn More

CD45RO Mouse anti-Human, PE-Cy5, Clone: UCHL1, BD

Mouse Monoclonal Antibody

550.00 CHF

Spezifikation

Antigen CD45RO
Klon UCHL1
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Konjugat PE-Cyanine5
Mehr Spezifikationen ansehen

Produkte 1
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15848218
Unterlagen ansehen Aktionsdetails
BD Biosciences
555494
100 Tests
550.00 CHF
1 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

The UCHL1 monoclonal antibody specifically binds to the 180 kDa isoform of CD45 (aka, the Leukocyte Common Antigen). CD45RO is a type I transmembrane glycoprotein that has cytoplasmic protein tyrosine phosphatase activity and functions in signal transduction pathways. This CD45 isoform does not include amino acid sequences encoded by the variable CD45 exons A, B, or C. CD45RO is expressed on most thymocytes, activated T cells, memory T cells, granulocytes and monocytes, but only on a proportion of resting T cells. CD45RO and CD45RA antibodies seem to define complementary, predominantly non-overlapping, populations in resting peripheral T cells, demonstrating heterogeneity within the CD8 and CD4 subpopulations. CD45RO binds to CD22.

Spezifikation

Spezifikation

CD45RO
Flow Cytometry
PE-Cyanine5
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
UCHL1
Monoclonal
Mouse
Human
Human IL-2-dependent T-cell line
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

For Research Use Only.