Learn More
CD45RB Monoclonal Antibody (PD7/26), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA512496
Additional Details : Gewicht : 0.01000kg
Beschreibung
MA5-12496 targets CD45RB in FACS, Western blot and IHC (P) applications and shows reactivity with Human and Mouse samples. This antibody is not recommended for BAF-3 cells in Western blot applications. The MA5-12496 immunogen is human peripheral blood lymphocytes maintained in T cell growth factor.
CD45RB is a receptor-type protein tyrosine phosphatase, CD45 glycoprotein. CD45 is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases, promotes cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45 isoforms differ in their extracellular domains, but share identical transmembrane and cytoplasmic domains. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45RB is expressed in microglia and inflammatory cells. In addition to involvement in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is involved in DNA fragmentation during apoptosis. CD45 is expressed on cells of the human hematopoietic lineage with the exception of mature red cells.Spezifikation
CD45RB | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P06800, P08575 | |
Ptprc | |
Human peripheral blood lymphocytes maintained in T cell growth factor | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
PD7/26 | |
Unconjugated | |
Ptprc | |
B220; CD45; CD45 antigen; CD45R; GP180; LCA; L-CA; Leukocyte common antigen; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Mouse | |
Protein G | |
RUO | |
19264, 5788 | |
4° C | |
Liquid |