missing translation for 'onlineSavingsMsg'
Learn More

CD45RA Monoclonal Antibody (MEM-56), PE-Cyanine5.5, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MHCD45RA18

448.02 CHF gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Additional Details : Gewicht : 0.02000kg

Artikelnummer. 10660284

  • 672.00 CHF / 1 Stück
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The R-phycoerythrin (PE)-Cy5.5® tandem conjugate permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser.

CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

CD45RA
Monoclonal
0.1mg/mL
Assay-dependent
PTPRC
B220; CD45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; LCA; L-CA; leucocyte common antigen; Leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; Lyt-4; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; PTPRC; Receptor-type tyrosine-protein phosphatase C; RT7; T100; T200; T200 glycoprotein; T200 leukocyte common antigen
Mouse, Mouse
purified
ASR
5788
0.1% Sodium azide
4° C, store in dark
IgG2b
Flow Cytometry, Flow Cytometry
MEM-56
PE-Cyanine5.5
PBS with 10% sucrose, BSA and 0.1% sodium azide
P08575
PTPRC
Human CD45RA
500 μL
Primary
1 Each
Human
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Analyte Specific Reagent. Analytical and performance characteristics are not established.