missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45R Monoclonal Antibody (RA3-6B2), APC-Cyanine7, Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen A15399
Additional Details : Gewicht : 0.00280kg
Beschreibung
CD45R Monoclonal antibody specifically detects CD45R in Feline, Human, Mouse samples. It is validated for Flow Cytometry.Spezifikation
CD45R | |
Monoclonal | |
0.1 mg/mL | |
PBS with proprietary stabilizer and <0.1% sodium azide | |
P06800, P08575 | |
PTPRC | |
exon A-specific epitope on CD45 from Abelson murine leukemia virus-induced pre-B cell lymphoma | |
100 μg | |
Primary | |
Feline, Human, Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
RA3-6B2 | |
APC-Cyanine7 | |
PTPRC | |
B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Rat | |
purified | |
RUO | |
101097123, 19264, 5788 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |