missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45R Monoclonal Antibody (RA3-6B2), APC-Cyanine5.5, Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen RM2619
Additional Details : Gewicht : 0.50000kg
Beschreibung
CD45R Monoclonal antibody specifically detects CD45R in Mouse samples. It is validated for Flow Cytometry.Spezifikation
CD45R | |
Monoclonal | |
APC-Cyanine5.5 | |
PTPRC | |
B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Rat | |
purified | |
RUO | |
19264 | |
4° C | |
Liquid |
Flow Cytometry | |
RA3-6B2 | |
PBS with 0.1% sodium azide | |
P06800 | |
PTPRC | |
exon A-specific epitope on CD45 | |
500 μL | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |