Learn More
CD45 Rat anti-Mouse, PE-Cy7, Clone: 30-F11, BD
Rat Monoclonal Antibody
227.00 CHF - 366.00 CHF
Spezifikation
Antigen | CD45 |
---|---|
Klon | 30-F11 |
Konzentration | 0.2mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15831419
|
BD Biosciences
561868 |
25 μg |
227.00 CHF
1 Stück |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15806878
|
BD Biosciences
552848 |
0.1 mg |
366.00 CHF
1 Stück |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The 30-F11 clone has been reported to react with all isoforms and both alloantigens of CD45, which is found on hematopoietic stem cells and all cells of hematopoietic origin, except erythrocytes. CD45 is a transmembrane glycoprotein which is expressed at high levels on the cell-surface, and its presence distinguishes leukocytes from non-hematopoietic cells. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family, where the intracellular carboxy-terminal region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated as A, B, and C, respectively). CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction and the CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific.
Spezifikation
CD45 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
IgG2b κ | |
Affinity Purified | |
Ptprc; CD45R; LCA; Leukocyte common antigen; T200; B220; Ly-5; Lyt-4 |
30-F11 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.