Learn More
CD45 Rabbit anti-Human, Unconjugated, Clone: 6O19, Proteintech
Artikelnummer. 17551496 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
20 μL
100 μL
Packungsgröße:
100 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17551496 20 μL 20 Mikroliter
17541496 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17551496 Lieferant Proteintech Lieferanten-Nr. 802971RR20UL
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Rabbit Recombinant Antibody
The CD45 antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human CD45. This antibody recognizes human, rat antigen. The CD45 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Spezifikation
Antigen CD45
Anwendungen Western Blot, Immunohistochemistry, Flow Cytometry
Klon 6O19
Konjugat Unconjugated
Wirtsspezies Rabbit
Menge 20 μL
Primär oder sekundär Primary
Gen-ID (Entrez) 5788
Zielspezies Human, Rat
Isotype IgG