Learn More
CD45 Mouse anti-Rat, FITC, Clone: OX-1, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 554877
Additional Details : Gewicht : 0.01000kg
Beschreibung
The OX-1 antibody reacts with all molecular forms of CD45 (Leukocyte Common Antigen) on all hematopoietic cells except erythrocytes. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Spezifikation
CD45 | |
Monoclonal | |
0.5mg/mL | |
Ptprc; Lca; Leucocyte common antigen; RT7; T200 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
OX-1 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Leukocyte Common Antigen-enriched Glycoprotein Fraction from Wistar Rat Thymocytes | |
0.5 mg | |
Immunology | |
Rat | |
IgG1 κ |
For Research Use Only.