missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Mouse anti-Human, BB515, Clone: HI30, BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 564585
Additional Details : Gewicht : 0.01000kg
Beschreibung
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow cytometry.
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow cytometry.Spezifikation
CD45 | |
Monoclonal | |
Brilliant Blue 515 (BB515) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Cell Biology | |
Human | |
Purified |
Flow Cytometry | |
HI30 | |
Vol. Per test: 5μL | |
PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5 | |
Human Peripheral Blood Leucocytes | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
IgG1 κ |
For Research Use Only.